Learn More
Brady™ BBP12 Label Printer
Product Code. 15512000
Click to view available options
Product Type:
Label Printer Kit EU
Label Printer with Unwinder
Label Printer with Unwinder, EU
Unit Size:
Each
Product Code. 15512000
Brand: Brady™ 195569
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Offers high print speed with the ability to handle a wide variety of identification labels. Brady™ BBP12 Label Printer is an entry level printer for laboratory sample identification.
  • Prints a wide range of labels from 10mm up to 112mm in width
  • Accurate image and barcode positioning on small labels
  • Standard 300 dpi print quality
  • Full colour LCD display
  • Comes with an external unwinder to print larger label rolls
  • Stand alone, as any universal keyboard can be connected
  • Dimensions (W x D x H): 202mm x 258mm x 173mm)
Specifications
Product Type Label Printer with Unwinder, EU
Display Type Color LCD
Print Technology Thermal Transfer
Width (Metric) 202 mm
Height (Metric) 173.00 mm
Depth (Metric) 258.00 mm
Dimensions (L x W) 173 x 202 mm
Color Capability Monochrome
Description BBP12 label printer with unwinder & Label-Mark6 Pro software, EU power supply
Includes This BBP12 label printer comes in a cardboard box. It ships with an unwinder, USB Cable, power cord, product CD, Quick Start Guide and a LabelMark Pro CD
Languages Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian (Bokmål), Polish, Portuguese, Russian, Slovak, Spanish, Swedish, Turkish
Memory 64 MB
Print Speed 100 labels/day
Resolution 300 dpi
Software Type LabelMark, Windows based driver for 3rd party software use, CodeSoft
Connector Types Serial, USB, Ethernet
Temperature Range (Metric) 5°C to 40°C
Weight (Metric) 2.5 kg
Certifications/Compliance UL; cUL; CE; RoHS; EAC
For Use With (Application) Asset identification, Barcode labelling, Wire and cable marking, Laboratory identification
Quantity 1 Ea.
Warranty 1 Year
Power Supply Details AC Adaptor
Voltage 110 to 240 V