Learn More
Brady™ BBP12 Label Printer
Product Code. 15430705 Shop All Brady Products
Change view
Click to view available options
Product Type:
Label Printer Kit EU
Label Printer with Unwinder
Label Printer with Unwinder, EU
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15430705 Label Printer with Unwinder Each
15512000 Label Printer with Unwinder, EU Each
15532000 Label Printer Kit EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15430705 Supplier Brady™ Supplier No. 195566
  • ONLINE EXCLUSIVE PRICE
    662.00€
    576.45€ / Pack of 1
    Save 85.55€
    12% Off
View all promotions
In Stock
Offers high print speed with the ability to handle a wide variety of identification labels. Brady™ BBP12 Label Printer is an entry level printer for laboratory sample identification.
  • Prints a wide range of labels from 10mm up to 112mm in width
  • Accurate image and barcode positioning on small labels
  • Standard 300 dpi print quality
  • Full colour LCD display
  • Comes with an external unwinder to print larger label rolls
  • Stand alone, as any universal keyboard can be connected
  • Dimensions (W x D x H): 202mm x 258mm x 173mm)
Specifications
Product Type Label Printer with Unwinder
Display Type Color LCD
Print Technology Thermal Transfer
Width (Metric) 202 mm
Height (Metric) 173.00 mm
Depth (Metric) 258.00 mm
Dimensions (L x W) 173 x 202 mm
Color Capability Monochrome
Description BBP12 label printer with unwinder, EU power supply
Includes This BBP12 label printer comes in a cardboard box. It ships with an unwinder, USB Cable, power cord, product CD and Quick Start Guide
Languages Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian (Bokmål), Polish, Portuguese, Russian, Slovak, Spanish, Swedish, Turkish
Memory 64 MB
Print Speed 100 labels/day
Resolution 300 dpi
Software Type LabelMark, Windows based driver for 3rd party software use, CodeSoft
Connector Types Serial, USB, Ethernet
Temperature Range (Metric) 5°C to 40°C
Weight (Metric) 2.5 kg
Certifications/Compliance UL; cUL; CE; RoHS; EAC
For Use With (Application) Asset identification, Barcode labelling, Wire and cable marking, Laboratory identification
Quantity 1 Ea.
Warranty 1 Year
Power Supply Details AC Adaptor
Voltage 110 to 240 V
Alternate Part Number BBP12-EU-UNWINDER