missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ CTA Medium

Product Code. 17372331
Click to view available options
Quantity:
8 mL
Media Type:
CTA Culture Medium with Dextrose
CTA Culture Medium with Lactose
CTA Culture Medium with Maltose
Packaging Type:
Tube
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17372331

Brand: BD 221633

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms; addition of carboyhydrates allows use in the determination of fermentation reactions

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type CTA Culture Medium with Dextrose
Formulation L-Cystine 0.5g/L, Pancreatic digest of Casein 20g/L, Agar 2.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Phenol red 0.017g/L
For Use With (Application) For maintaining the growth of microorganisms
Packaging Type Tube
Quantity 8 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.