Learn More
R&D Systems™ BAY 9835
Description
BAY 9835 is a potent ADAMTS7 and ADAMTS12 inhibitor (IC50 values are 6 nM and 30 nM, respectively). BAY 9835 is selective over other metalloproteases (ADAMTS4, ADAMTS5, ADAM8, ADAM10, ADAM17, MMP2, MMP12, MMP14, MMP15). BAY 9835 is the first orally bioavailable ADAMTS7 inhibitor.
Specifications
Specifications
| Chemical Name or Material | 5,6-Difluoro-N-[[(4S)-4-(1-methyl-1H-pyrazol-5-yl)-2,5-dioxo-4-imidazolidinyl]methyl]-4'-(trifluoromethyl)-[1,1'-biphenyl]-2-carboxamide |
| CAS | 3032487-31-0 |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Synthetic Metalloprotease Inhibitors |
| Molecular Formula | C22H16F5N5O3 |
| Product Type | Bioactive Small Molecule |
| Purity | 98% |