missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 805
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BAY 805 is a potent and selective inhibitor of USP21 (IC50 = 6 nM, Kd = 2.2 nM). BAY 805 induces NF-κB activation (EC50= 17 nM in a cell-based reporter assay).
Specifications
Specifications
| Chemical Name or Material | N-[(1R)-1-[[[5-[(4-Cyanophenyl)methyl]-1,3,4-thiadiazol-2-yl]amino]carbonyl]-3-methylbutyl]-1-(trifluoromethyl)cyclohexanecarboxamide |
| CAS | 2925481-88-3 |
| Quantity | 50 mg |
| Target | Deubiquitinating Enzyme Inhibitors |
| Molecular Formula | C24H28F3N5O2S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction