Learn More
Dräger™ Battery/Charging Accessories - Multinational Power Supply
Product Code. 10312859
Change view
Click to view available options
Type:
Power Supply
Single Charger
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10312859 Power Supply Each
15666544 Single Charger Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10312859 Supplier Dräger™ Supplier No. 8315805
  • ONLINE EXCLUSIVE PRICE
    446.00€
    366.47€ / Pack of 1
    Save 79.53€
    17% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Specifications
Dimensions (L x W x H) 6 x 6 x 6 in. (15.24 x 15.24 x 15.24cm)
Type Power Supply
Height (English) 6 in.
Height (Metric) 15.24 cm
Length (English) 6 in.
Length (Metric) 15.24 cm
Voltage 100/240VAC
Width (English) 6 in.
Width (Metric) 15.24 cm