Learn More
Testo™ Batteries
Click to view available options
For Use With (Equipment):
Testo 175-T1, 175-T2, 177
Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2
Testo 175-T3/175-H1/175-H2/S1
Testostor 171
Unit Size:
Each
Description
- 3.6V
- 0.8Ah
- ½AA
Specifications
Specifications
| Product Type | Battery |
| For Use With (Equipment) | Testo 175-T3/175-H1/175-H2/S1 |