Learn More
Testo™ Batteries
Product Code. 10192492
Change view
Click to view available options
For Use With (Equipment):
Testo 175-T1, 175-T2, 177
Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2
Testo 175-T3/175-H1/175-H2/S1
Testostor 171
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10192492 Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2 Each
10002772 Testo 175-T1, 175-T2, 177 Each
10105991 Testostor 171 Each
11881724 Testo 175-T3/175-H1/175-H2/S1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10192492 Supplier Testo™ Supplier No. 0515 0175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this battery with a variety of products. Testo™ Batteries are compatible with Testo data loggers.
  • 3.6V
  • 0.8Ah
  • ½AA
Specifications
Product Type Battery
For Use With (Equipment) Testo 175-T3, 175-H1, 175-H2, 175-S1, 175-S2
Voltage 3.6 V
Amperage 0.8 Ah