Learn More
Hirschmann™ Batch Calibration Certificate
Description
- Quality certificate shows the same batch number
- Certificate provides information on the mean value of the production lot, the standard deviation and the prescribed tolerance
- Accessory item
Specifications
Specifications
| For Use With (Equipment) | Measuring Instruments |
| Certifications/Compliance | Batch Calibration |
| Product Type | Certificate |