Learn More
Memmert™ Basic Series Water Baths
Product Code. 10329342 Shop All Memmert Products
Click to view available options
Capacity (Metric):
10 L
14 L
22 L
29 L
45 L
Unit Size:
Each
Product Code. 10329342
Brand: Memmert™ WNB10
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Controller Type Microprocessor PID
Capacity (Metric) 10 L
Certifications/Compliance CE, VDE, GS
Display Digital LED
For Use With (Application) For research laboratories, food and beverage, temperature control
Material Stainless Steel
Model WNB 10
Temperature (Metric) Maximum 95°C
Temperature Resolution ±0.1°C
Length (Metric) 578 mm
Length (Metric) Interior 350 mm
Width (Metric) 356 mm
Width (Metric) Interior 210 mm
Height (Metric) 238 mm
Height (Metric) Interior 140 mm
Depth (Metric) Interior 350 mm
Shipping Weight (Metric) 17 kg
Weight (Metric) 13 kg
Voltage 230 V
Wattage 1.2 kW
Frequency 50/60 Hz