Learn More
Basic Red 12 (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Basic Red 12 (Technical Grade) |
| CAS | 6320-14-5 |
| Molecular Formula | C25H29N2 . Cl |
| Quantity | 10 mg |
| Synonym | Astraphloxin |
| InChI Formula | InChI=1S/C25H29N2.ClH/c1-24(2)18-12-7-9-14-20(18)26(5)22(24)16-11-17-23-25(3,4)19-13-8-10-15-21(19)27(23)6;/h7-17H,1-6H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CN1C(=CC=CC2=[N+](C)c3ccccc3C2(C)C)C(C)(C)c4ccccc14 |
| IUPAC Name | 1,3,3-trimethyl-2-[3-(1,3,3-trimethylindol-1-ium-2-yl)prop-2-enylidene]indole;chloride |
| Molecular Weight (g/mol) | 393 |
| Formula Weight | 392.2019 |