Learn More
Eppendorf™ White Glove Delivery per unit
Product Code. 15793685
Change view
Click to view available options
Product Type:
White Glove Delivery (>180 kg)
White Glove Delivery (<100 kg)
White Glove Delivery (100 to 180 kg)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15793685 White Glove Delivery (>180 kg) Each
15773685 White Glove Delivery (<100 kg) Each
15783685 White Glove Delivery (100 to 180 kg) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15793685 Supplier Eppendorf™ Supplier No. 2277000004
 Restricted Item
Estimated Shipment: 23-12-2026
Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging. Shipment with stair climb to be ordered when needed.
  • White glove delivery per unit
  • Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging
  • Shipment with stair climb to be ordered when needed
TRUSTED_SUSTAINABILITY
Specifications
Product Type White Glove Delivery (>180 kg)