Learn More
Ansell Edmont™ Barrier™ Disposable Hand Specific Nitrile Gloves
Manufactured from five layer film for wide spectrum chemical resistance. Ansell™ Barrier™ Disposable Hand Specific Nitrile Gloves can also be used as liners for more dangerous applications.
Brand:  Ansell Edmont™ 02-100_8
Product Code. 11711573
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Chemical resistant film gloves with an ergonomic design
  • Integral, non-woven liner absorbs and dissipates perspiration
  • Patented seam technology locks out chemicals and keeps the protective film and liner together as a single unit
  • Worn alone, Barrier™ gloves provide flexibility and tactility
  • Where the risk of physical damage is greater, the gloves should be worn as liners under heavier nitrile, neoprene, or PVC gloves when used for riskier applications where risk of phyiscal damage is greater