Learn More
Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories
Product Code. 12760696 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Organic Removal Cartridge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12760696 Organic Removal Cartridge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12760696 Supplier Thermo Scientific™ Supplier No. D440265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure™ Glass Still, including storage bottles and deionization cartridges.
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure Glass Still. Accessories include dual solenoid, deionization cartridges, storage solutions, single cartridge deionizer, dual cartridge deionizer, deionizer accessories and cartridges, storage bottles and automatic collection system.
TRUSTED_SUSTAINABILITY
Specifications
Description Organic Removal Cartridge
For Use With (Equipment) D2 (organic and chlorine removal)
Type Cartridge, Organic Removal