Learn More
Equip Labo™ Bare Benches
Product Code. 12658832
Change view
Click to view available options
Depth (Metric):
600 mm
750 mm
Width (Metric):
600 mm
900 mm
120 cm
150 cm
180 cm
Top Style:
Stretched Sandstone
White Extruded Stoneware
White Melamine
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Width (Metric) Top Style unitSize
12658832 600 mm 600 mm White Melamine Each
12567716 600 mm 120 cm White Melamine Each
12516496 600 mm 150 cm White Extruded Stoneware Each
12536486 600 mm 180 cm White Melamine Each
12579026 750 mm 900 mm White Melamine Each
12527806 750 mm 900 mm White Extruded Stoneware Each
12625592 750 mm 120 cm White Melamine Each
12536696 750 mm 120 cm Stretched Sandstone Each
12615592 750 mm 150 cm White Melamine Each
12678832 750 mm 180 cm White Melamine Each
12546486 750 mm 600 mm White Melamine Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 12658832 Supplier Equip Labo™ Supplier No. 8003
  • ONLINE EXCLUSIVE PRICE
    487.00€
    407.20€ / Pack of 1
    Save 79.80€
    16% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Specifications
Product Type Bench
Depth (Metric) 600 mm
Load Bearing Capacity (Metric) 130 kg
Top Style White Melamine
Warranty 12 Months
Width (Metric) 600 mm