Learn More
Equip Labo™ Bare Benches
Product Code. 12579026
Click to view available options
Depth (Metric):
600 mm
750 mm
Width (Metric):
600 mm
900 mm
120 cm
150 cm
180 cm
Top Style:
Stretched Sandstone
White Extruded Stoneware
White Melamine
Unit Size:
Each
Product Code. 12579026
Brand: Equip Labo™ 8063
Specifications
Product Type Bench
Depth (Metric) Shelf 150 mm
Depth (Metric) 750 mm
Dimensions (D x W x H) 750 x 900 x 900 mm
Height (Metric) 900 mm
Load Bearing Capacity (Metric) 130 kg
Top Style White Melamine
Warranty 12 Months
Width (Metric) 900 mm