Learn More
Equip Labo™ Bare Benches
Brand:  Equip Labo™ 8067
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12527806
  • 895.00€ / Each
Estimated Shipment: 06-06-2024
Specifications
750 x 900 x 900 mm
12 Months
900 mm
130 kg
150 mm
White Extruded Stoneware
Bench
750 mm
900 mm