Learn More
Corning™ Bar-coded Cryogenic Vials
Product Code. 15515729 Shop All Corning Products
Change view
Click to view available options
Barcode:
1D
2D
Thread Style:
External
External threaded
Internal
Unit Size:
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Barcode Thread Style Unit Size
15515729 1D Internal Pack of 500
15595719 2D Internal Pack of 500
15505729 2D External threaded Pack of 500
15545729 1D External Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15515729 Supplier Corning™ Supplier No. 8672
  • ONLINE EXCLUSIVE PRICE
    413.00€
    385.31€ / Pack of 500
    Save 27.69€
    6% Off
Estimated Shipment: 04-02-2027
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
  • Manufactured from polypropylene
  • Leak proof seal for integrity of storage
  • Autoclavable vials and caps, can be re-sterilized
  • Unique 1D, 2D bar-codes and human readable characters
  • Features star shaped cavity in cap and is compatible with decapping equipment
  • Locking base stabilizes tubes for automated handling
  • Graduations and writing patch for manual use also
  • Each tube is checked for clarity and readability of 2D code
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Orange
Color Natural
Sterility Sterile
Volume (Metric) 2 mL
For Use With (Application) Cell biology, cell banking, compound cryopreservation
No. per Case 500
No. per Pack 50
Thread Style Internal
Base Style Round, Self-standing
Barcode 1D
Closure Material Polyethylene