Learn More
Cole-Parmer™ Barbed T Connector
Product Code. p-7116320
Click to view available options
Hose Barb:
1/8 in.
3/32 in.
Unit Size:
Pack of 100
Product Code. 11769709
Brand: Cole-Parmer™ 0636580
Only null left
Add to basket
Add to basket
For use with a variety of applications and tubing
  • Commercial grade barbed reducing straight connector
  • Polypropylene construction
  • Available in a avriety of sizes
  • Pack of 100
Specifications
Material Polypropylene
Packaging Quantity 100 Connectors
Hose Barb 1/8 in.
Color Natural
Product Type Commercial Grade T Connector