Learn More
DiscoverX BAM (8-22)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
BAM (8-22) is a opioid peptide fragment of the bovine adrenal medulla (BAM22) neuropeptide that is involved in inflammation, pain, and morphine tolerance. Induces itch and nociceptive sensations. Lacks the met-enkephalin motif of BAM22, hence lacking affinity for opioid receptors. Selective agonist of the sensory neuron specific receptors SNSR3 and SNSR4. Provided as 1mg of dry powder.
Specifications
Specifications
| Product Type | Control Ligands |
| Quantity | 1 mg |