Learn More
DiscoverX BAM (8-22)
Product Code. 17312539
Encompass
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 17312539
Brand: DiscoverX 921215
BAM (8-22) is an agonist for MRGPRX1. Provided as 1mg of dry powder.
BAM (8-22) is a opioid peptide fragment of the bovine adrenal medulla (BAM22) neuropeptide that is involved in inflammation, pain, and morphine tolerance. Induces itch and nociceptive sensations. Lacks the met-enkephalin motif of BAM22, hence lacking affinity for opioid receptors. Selective agonist of the sensory neuron specific receptors SNSR3 and SNSR4. Provided as 1mg of dry powder.
Specifications
Product Type Control Ligands
Quantity 1 mg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.