Learn More
Lenz Laborglasintrumente Ball Members Inch Range
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01211909
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 10050341
  • 89.04€ / Pack of 10
Estimated Shipment: 19-09-2024
Description
  • Ball members
  • Inch range
  • Acc. to DIN 12264
  • Made of DURAN™ tubing
Specifications
Specifications
S 19
Duran Glass