Learn More
Cole-Parmer™ Essentials Hollow Polypropylene (PP) Ball Bath Covers
Product Code. 15398060 Shop All Cole-Parmer Products
Click to view available options
For Use With (Equipment):
Water Baths
Unit Size:
Pack of 1000
Product Code. 15398060
Brand: Cole-Parmer™ Essentials 0682104
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Hollow Polypropylene (PP) Ball Bath Covers reduce vapor and heat loss by 75% with one layer of balls.
  • Polypropylene balls form a tight cover over liquids to prevent change in temperature and reduce evaporation.
  • Use instead of a bath lid.
Specifications
For Use With (Equipment) Water Baths
Type Ball Bath Cover
Material Polypropylene
Quantity 1000 Pk.
Diameter (Metric) Min. Opening 20 mm