Learn More
Sartorius Balance and Scale Weighing Pans
Product Code. p-4235126 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Cubis 10mg and 100mg Balances
Unit Size:
Each
Product Code. 12837232
Brand: Sartorius YWP03MS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
To prevent any damage to the surface of weights, Sartorius offers you a wide range of accessories for the proper handling, storage and cleaning of weights.
Weighing pans for use with Sartorius Balances.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Bottom Shape Flat
Color Silver
Disposable Reusable
Material Stainless Steel
Shape Rectangular
Sterility Non-sterile
Type Weighing Pan
For Use With (Application) Weighing samples
For Use With (Equipment) Cubis 10mg and 100mg Balances