Learn More
Sartorius Balance Printers
GREENER_CHOICE
Product Code. 10506089 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
CPA Balance, Extended Balance, Moisture Analyzers
CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.)
Dimensions (L x W x H):
215 x 155 x 78 mm
230 x 158 x 80 mm
Unit Size:
Each
Product Code. 10506089
Brand: Sartorius YDP100CE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Transfer data automatically or manually. Sartorius Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
  • Built-in real-time clock
  • Built-in statistics program
  • Counter for consecutive numbering of transactions can be activated
  • High contrast LCD shows date or time as well as current transaction number
  • Integrated rechargeable batteries with battery level indicator
  • Ultrafast print head
  • Runs on the balance's power; no separate power supply necessary
  • Easy to change paper roll and ink ribbon cassette
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.)
Dimensions (L x W x H) 230 x 158 x 80 mm
Length (Metric) 230 mm
Width (Metric) 158 mm
Height (Metric) 80 mm
Includes Interface cable, Paper roll, 4 rechargeable batteries
Weight (Metric) 0.77 kg
Type Data Printer