Learn More
Avantor™ BAKERBOND Speedisk™ Oil and Grease Extraction Disk
Product Code. 15578464 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15578464 20/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15578464 Supplier Avantor™ Supplier No. 806006
  • ONLINE EXCLUSIVE PRICE
    631.55€
    534.97€ / Pack of 1
    Save 96.58€
    15% Off
 Restricted Item
Estimated Shipment: 25-03-2027
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Oil and Grease Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) For Use with Slightly Polar to Non-Polar Hydrocarbons, EPA Method 1664, Rev. A.
For Use With (Equipment) Expanded Extraction Station (8095-06)
Type Oil and Grease Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm