Learn More
J.T. Baker BAKERBOND spe™ Silica Gel (SiOH) Disposable Extraction Columns, J.T. Baker™
Product Code. 10781272 Shop All J.T.Baker Products
Click to view available options
Bed Weight:
500 mg
1000 mg
Column Capacity (Metric):
3 mL
6 mL
Quantity:
Pack of 250
Pack of 30
Pack of 50
Unit Size:
Each
Pack of 30
Pack of 50
Product Code. 10781272
Brand: J.T. Baker 708607
Specifications
Column Capacity (Metric) 6 mL
Column Phase Silica gel
Description Disposable extraction column
Bed Weight 1000 mg
Format Granules
Particle Size 40 μm
Sorbent Silica
Pore Size 60 angstroms
Quantity Pack of 30
Color White