Learn More
J.T. Baker BAKERBOND spe™ Silica Gel (SiOH) Disposable Extraction Columns, J.T. Baker™
Product Code. 10381511 Shop All J.T.Baker Products
Change view
Click to view available options
Bed Weight:
500 mg
1000 mg
Column Capacity (Metric):
3 mL
6 mL
Quantity:
Pack of 250
Pack of 30
Pack of 50
Unit Size:
Each
Pack of 30
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Column Capacity (Metric) Quantity unitSize
10381511 500 mg 3 mL Pack of 50 Pack of 50
10474911 500 mg 6 mL Pack of 30 Pack of 30
10781272 1000 mg 6 mL Pack of 30 Pack of 30
15547654 500 mg 6 mL Pack of 250 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10381511 Supplier J.T. Baker Supplier No. 708603
  • ONLINE EXCLUSIVE PRICE
    367.00€
    310.43€ / Pack of 50
    Save 56.57€
    15% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Specifications
Column Capacity (Metric) 3 mL
Column Phase Silica gel
Description Disposable extraction column
Bed Weight 500 mg
Format Granules
Particle Size 40 μm
Sorbent Silica
Pore Size 60 angstroms
Quantity Pack of 50
Color White