Learn More
Thermo Scientific™ Oxoid™ Baird-Parker Agar Base (Dehydrated)
Product Code. 12754156
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
Product Code. 12754156
Brand: Thermo Scientific™ CM0275T
Isolate and enumerate Staphylococcus aureus with this selective and sensitive medium.
Isolate and enumerate Staphylococcus aureus with selective and sensitive Thermo Scientific™ Baird-Parker Agar Base (Dehydrated).
Baird Parker Agar Base contains carefully balanced selective inhibitory agents, glycine, lithium, and tellurite, to suppress the growth of most of the bacteria present in foods without inhibiting coagulase-positive staphylococci. The presence of egg yolk emulsion and sodium pyruvate aid the recovery of damaged cells for presumptive identification of Staphylococcus aureus in food specimens.
Baird-Parker Agar Base is widely recommended by national and international bodies for the isolation of Staphylococcus aureus1.
  • Easy interpretation of test results as Staphylococcus aureus forms grey-black shiny colonies with surrounding clear zones.
  • Greater recovery of damaged staphylococci than on other media2.
Baird-Parker2 developed this medium from the tellurite-glycine formulation of Zebovitz et al.3 and improved its reliability in isolating Staphylococcus aureus from foods. Baird-Parker added sodium pyruvate, to protect damaged cells and aid their recovery3
Egg yolk emulsion makes the medium yellow and opaque. Staphylococcus aureus reduces tellurite to form grey-black shiny colonies and then produces clear zones around the colonies by proteolytic action. This clear zone with typical grey-black colony is diagnostic for Staphylococcus aureus. On further incubation, most strains of Staphylococcus aureus form opaque haloes around the colonies.
Some strains of Staphylococcus saprophyticus may also produce both clear zones and opaque haloes but can be distinguished by the longer incubation required4.
Colonies typical of Staphylococcus aureus, but without an egg yolk reaction, should also be tested for coagulase production5. Egg yolk reaction negative Staphylococcus aureus may occur in some food, especially cheese.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Appearance Straw powder
For Use With (Equipment) Egg Yolk Tellurite Emulsion
pH 6.6 to 7.0
Quantity 5 kg
For Laboratory Use Only