Learn More
Gibco™ Bacto™ Proteose Peptone No. 3
Product Code. 16239751
Change view
Click to view available options
Quantity:
2 kg
50 kg
Unit Size:
2kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16239751 50 kg 50kg
16289771 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16239751 Supplier Gibco™ Supplier No. 211692
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media.
  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Microbiological and mammalian cell culture media
Product Line Bacto
Quantity 50 kg
Source Animal Origin