Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Product Code. p-4525702
Click to view available options
Packaging:
500/Pk.
Swab Configuration:
Single Applicator
Unit Size:
Pack of 500
Product Code. 11992642
Brand: Thermo Scientific™ R723000
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Blue cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
TRUSTED_SUSTAINABILITY
Specifications
Media Type Amies Clear
Swab Configuration Single Applicator
Closure Color Blue
Description Amies Clear
Packaging 500/Pk.
Quantity 1 Tube/Envelope
Shaft Material Plastic