Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Description
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
- Blue cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Specifications
| Media Type | Amies Clear |
| Swab Configuration | Single Applicator |
| Closure Color | Blue |
| Description | Amies Clear |
| Packaging | 500/Pk. |
| Quantity | 1 Tube/Envelope |
| Shaft Material | Plastic |