Learn More
Thermo Scientific™ BactiDisks™ Candida tropicalis ATCC™ 13803™
Description
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
Specifications
Specifications
| ATCC® | 13803TM* |
| Description | Candida tropicalis |
| Format | BactiDisks |
| Product Type | Bactidisk |
| Target Organism Class | Fungi |
| Test Type | Qualitative QC |
| Quantity | 10 Disk/Vial |
| Strain Designations | FDA PCI M-59 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.