Learn More
Novacyt - Microgen Bioproducts Bacterial ID System Reagent/Consumable
Product Code. 15967122 Shop All Novacyt Group Products
Click to view available options
Product Type:
Indole Kovacs Reagent
Mineral Oil
Ninhydrin-filled Vial
Nitrate A Reagent
Nitrate B Reagent
Oxidase Strips
PYR Reagent
TDA Reagent
VP I Reagent
VP II Reagent
Unit Size:
10mL
50 strips
50mL
Product Code. 15967122
Brand: Novacyt - Microgen Bioproducts MID61G
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Additional/supplemental reagent/consumable for biochemical identification system.
  • Replacement/supplemental reagent/consumable.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
Product Type Oxidase Strips
For Use With (Application) Biochemical Identification Kits
compliance-icons
Product Identifier
  • MID-61G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : oxidase strips