Learn More
Novacyt - Microgen Bioproducts Bacterial ID System Reagent/Consumable
Product Code. 15957122 Shop All Novacyt Group Products
Click to view available options
Product Type:
Indole Kovacs Reagent
Mineral Oil
Ninhydrin-filled Vial
Nitrate A Reagent
Nitrate B Reagent
Oxidase Strips
PYR Reagent
TDA Reagent
VP I Reagent
VP II Reagent
Unit Size:
10mL
50 strips
50mL
Product Code. 15957122
Brand: Novacyt - Microgen Bioproducts MID61F
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Additional/supplemental reagent/consumable for biochemical identification system.
  • Replacement/supplemental reagent/consumable.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
Product Type Indole Kovacs Reagent
For Use With (Application) Biochemical Identification Kits
compliance-icons compliance-icons compliance-icons
Product Identifier
  • MID-61F
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Supplemental information
  • MIXTURE LIST-Contains : Iso-amyl alcohol, Hydrochloric Acid