Learn More
Novacyt - Microgen Bioproducts Bacillus ID
Product Code. 17323221 Shop All Novacyt Group Products
Ansicht ändern
Click to view available options
No. of Reactions:
12 per test strip
Type:
Test Strip
Packungsgröße:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. No. of Reactions Type Packungsgröße
17323221 12 per test strip Test Strip 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17323221 Lieferant Novacyt - Microgen Bioproducts Lieferanten-Nr. MID66
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Microgen Bioprpducts Bacillus ID. For the identification of mesophilic Bacillus species isolated from food materials and the environment.
  • Microgen Bioproducts Bacillus-ID has been developed for the identification of mesophilic Bacillus spp. and related genera isolated from food, food ingredients and beverages either as food spoilage or food poisoning organisms.
Spezifikation
Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Arabinose, Cellobiose, Inositol, Mannitol, Mannose, Raffinose, Rhamnose, Salicin, Sorbitol, Sucrose, Trehalose, Xylose, Adonitol, Galactose, M-D-Mannopyranoside, M-D-Glucopyranoside, Inulin, Melezitose, Indole, Nitrate, Arginine, Citrate, Voges Proskauer
Format 20 x 3 mL bottles, 20 Test strips
Sample Type Bacterial Isolate
Sensitivity 0.98
Type Test Strip
For Use With (Application) An Identification System for The Mesophilic Bacillus Species and Genera From Food and Clinical Sources
Includes 20 Test strips
No. of Reactions 12 per test strip
Organism Group Monera
Packaging Type Carton with pouches and strip holding tray
Quantity 20 x 3 mL bottles, 20 Test strips
Species Mesophilic Bacillus species and Genera from Food and Clinical sources
Specificity 0.98
Target Organism Class Bacteria
Test Time Read after 24 hours and 48 hours incubation
Mehr anzeigen Weniger anzeigen