missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ B3GNT5 Recombinant Protein

Brand:  Abnova™ H00084002-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115583

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH28058
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
67.32
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B3GNT5
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
84002
B3GNT5 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
B3GN-T5/beta3Gn-T5
B3GNT5
Wheat Germ (in vitro)
GST
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions