Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Product Code. 30041440
Click to view available options
Dimensions (LxW):
12.70 mm (Dia)
12.70 mm (W) x 25.40 mm (H)
20.32 mm (W) x 36.50 mm (H)
25.40 mm (W) x 12.45 mm (H)
25.40 mm (W) x 36.50 mm (H)
25.40 mm (W) x 66.68 mm (H)
9.53 mm (Dia)
Color:
White
Yellow
Shape:
Rectangle
Round
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2500
Pack of 3000
Product Code. 30041440
Brand: Brady™ 361694
Only null left
Add to basket
Add to basket
Nylon cloth labels for BBP33/i3300 printers.
For BBP33/i3300 printers.
Specifications
Certifications/Compliance UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material Nylon Cloth (B-499)
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling, Electrical labelling, General identification, Laboratory labelling, Warehouse marking, Wire and cable labelling
For Use With (Equipment) BBP33, i3300, i5300
Labels per Row 1
Shape Rectangle
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0065 in.
Thickness (Metric) 0.166 mm
Vertical Repeat (Metric) 15.40 mm
Dimensions (LxW) 25.40 mm (W) x 12.45 mm (H)
Length (English) 12.45 mm
Width (English) 25.40 mm
Width (Metric) 25.40 mm
Width (Metric) Printable Area 30.48 mm
Height (English) 0.49 in.
Height (Metric) 12.45 mm
Alternate Part Number B33-659-499