Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Product Code. 30041407
Change view
Click to view available options
Dimensions (LxW):
12.70 mm (Dia)
12.70 mm (W) x 25.40 mm (H)
20.32 mm (W) x 36.50 mm (H)
25.40 mm (W) x 12.45 mm (H)
25.40 mm (W) x 36.50 mm (H)
25.40 mm (W) x 66.68 mm (H)
9.53 mm (Dia)
Color:
White
Yellow
Shape:
Rectangle
Round
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2500
Pack of 3000
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Color Shape unitSize
30041407 12.70 mm (W) x 25.40 mm (H) White Rectangle Pack of 1000
30041429 12.70 mm (Dia) Yellow Round Pack of 3000
30041441 12.70 mm (Dia) White Round Pack of 3000
30041459 20.32 mm (W) x 36.50 mm (H) White Rectangle Pack of 2500
30041401 9.53 mm (Dia) White Round Pack of 3000
30041423 9.53 mm (Dia) Yellow Round Pack of 3000
30041445 25.40 mm (W) x 66.68 mm (H) White Rectangle Pack of 1500
30041408 25.40 mm (W) x 36.50 mm (H) White Rectangle Pack of 3000
30041440 25.40 mm (W) x 12.45 mm (H) White Rectangle Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30041407 missing translation for 'mfr' Brady™ missing translation for 'supplierNo' 361679
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Nylon cloth labels for BBP33/i3300 printers.
For BBP33/i3300 printers.
Especificaciones
Certifications/Compliance UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material Nylon Cloth
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling, Electrical labelling, General identification, Laboratory labelling, Warehouse marking, Wire and cable labelling
For Use With (Equipment) BBP33, i3300, i5300
Labels per Row 1
Shape Rectangle
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0065 in.
Thickness (Metric) 0.166 mm
Vertical Repeat (Metric) 28.40 mm
Dimensions (LxW) 12.70 mm (W) x 25.40 mm (H)
Length (English) 25.40 mm
Width (English) 12.70 mm
Width (Metric) 12.70 mm
Width (Metric) Printable Area 25.40 mm
Height (English) 1 in.
Height (Metric) 25.40 mm
Alternate Part Number B33-620-499
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.