Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Click to view available options
Dimensions (LxW):
12.70 mm (Dia)
12.70 mm (W) x 25.40 mm (H)
20.32 mm (W) x 36.50 mm (H)
25.40 mm (W) x 12.45 mm (H)
25.40 mm (W) x 36.50 mm (H)
25.40 mm (W) x 66.68 mm (H)
9.53 mm (Dia)
Color:
White
Yellow
Shape:
Rectangle
Round
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2500
Pack of 3000
Description
For BBP33/i3300 printers.
Specifications
Specifications
| Certifications/Compliance | UL Recognized,CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only |
| Description | Nylon Cloth Labels for BBP33/i3300 Printers |
| Adhesive Type | Acrylic, Permanent |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Nylon Cloth |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Component and Equipment Labelling, Electrical Labelling, General Identification, Laboratory Labelling, Warehouse Marking, Wire and Cable Labelling |
| Show More |