Learn More
B02, Tocris Bioscience™
RAD51 recombinase inhibitor
Brand:  Tocris Bioscience 6392/5
Encompass
Product Code. 15917940
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
B02 is a RAD51 recombinase inhibitor, exhibiting selectivity for human RAD51 over the E.coli homologue RecA (IC50 values are 27.4 μM and >250 μM, respectively). Inhibits homologous recombination and increases cellular sensitivity to DNA damage in vitro. Enhances the therapeutic effect of cisplatin (Cat. No. 2251) on tumor cells in vivo.
Specifications
Specifications
1290541-46-6
3-(Phenylmethyl)-2-[(1E)-2-(3-pyridinyl)ethenyl]-4(3H)-quinazolinone
5 mg