Learn More
Alpha Packaging™ B-safe™ Polyethylene Biohazard Bag
Product Code. 12320609
Change view
Click to view available options
Height (Metric):
1015 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12320609 1015 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12320609 Supplier Alpha Packaging™ Supplier No. 12320609
  • ONLINE EXCLUSIVE PRICE
    69.80€
    63.10€ / Pack of 25
    Save 6.70€
    9% Off
Estimated Shipment: 16-12-2026
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
  • Comes in loose packs or benchtop dispenser boxes.
  • For heavy-duty disposal
  • Marked on both sides with biohazard symbol and warnings:
    • "Biohazard, Danger of Infection"
    • "Burn Without Opening"
  • Warning in English, French, and German
  • Polyethylene (500 gauge)
Specifications
Color Yellow
Disposable Single-use
Height (Metric) 1015 mm
Length (Metric) 790 mm
Length (English) 31 in.
Height (English) 39.9 in.
Material Polyethylene
Product Type Biohazard Bag