Learn More
Thermo Scientific™ B-PER™ Bacterial Protein Extraction Reagent
Code produit. p-4531883 Tous les produits Thermo Scientific Produits
Click to view available options
Quantity:
165 mL
250 mL
500 mL
Conditionnement:
165mL
250mL
500mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11834091
Marque: Thermo Scientific™ 78243
Edge
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.
Perform efficient, gentle lysis and extraction of E.coli and other bacterial cells with Thermo Scientific™ B-PER Bacterial Protein Extraction Reagents.
These easy-to-use cell lysis reagents are nonionic detergent solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. Certain formulations include DNase I and lysozyme enzyme supplements to improve yield of large molecular weight proteins that are difficult to purify. B-PER Reagents are compatible with the addition of standard protease inhibitors and with downstream protein purification methods, such as GST purification and His-tagged immobilized metal affinity chromatography (IMAC).
Highlights:
  • Ready to use – B-PER Reagents provide one-step E.coli cell lysis by a mild, nonionic detergent (proprietary) in your choice of Tris or phosphate buffer formulations
  • Fast and simple – just add B-PER Reagent to a bacterial pellet, shake for 10 minutes and recover soluble proteins after pelleting the cell debris
  • Excellent yields – recover both soluble and insoluble recombinant protein from bacterial lysates and purify inclusion bodies to near-homogeneous levels
  • Flexible – B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
  • Compatible – completely compatible with addition of protease inhibitor cocktails, and resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications
Convenient, ready-to-use formats:
  • B-PER and B-PER II (2X) Reagents are free of enzymatic components
  • B-PER Reagent (in phosphate buffer) is an amine-free formulation for direct compatibility with labeling and crosslinking
  • B-PER with Enzymes and B-PER Direct with Enzymes include lysozyme and DNase I for improved lysis and recovery
Includes:
Reagent packages are available in Tris and phosphate buffer formulations; Kits contain Tris-based B-PER Reagent plus lysozyme and DNase I enzymes.
TRUSTED_SUSTAINABILITY
Spécification
Description B-PER Bacterial Protein Extraction Reagent
Quantity 165 mL
Product Type Bacterial Protein Extraction Reagent
Volume (Metric) 165 mL
Content And Storage Store at room temperature.
Product Line B-PER
For Research Use Only. Not for use in diagnostic procedures.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis