Learn More
Tocris Bioscience™ B I09
GREENER_CHOICE
Product Code. 18705909
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18705909
Brand: Tocris Bioscience™ 6009/10
IRE1 endonuclease inhibitor; cell permeable
B I09 is an IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1607803-67-7
Quantity 10 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C16H17NO5
Formula Weight 303.31g/mol
Color Beige
Product Type Inhibitor