Learn More
Brady™ B-427 Self-laminating Vinyl Wire Marker Labels
Product Code. 11543075
Click to view available options
Height (Metric):
36.5 mm
57.2 mm
95.3 mm
Labels per Row:
1
2
3
Vertical Repeat (Metric):
30.2 mm
41.3 mm
60.3 mm
101.6 mm
Unit Size:
1 pack
1 roll
Product Code. 11543075
Brand: Brady™ 072011
Only null left
Add to basket
Add to basket
Vinyl self-laminating labels for wire and electrical labeling.
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
  • For matte finish
Specifications
Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color Yellow, Transparent
Finish Matte
Format Die-cut
Material Self-laminating Vinyl
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
For Use With (Equipment) BBP11;BBP12;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Horizontal Repeat (Metric) 27.31 mm
Labels per Row 3
Quantity 3000 Labels
Shape Rectangle
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 101.6 mm
Wire Gauge 1 AWG to 400 MCM
Dimensions (LxW) 25.40 mm (W) x 95.25 mm (H)
Width (English) 1 in
Width (Metric) 25.4 mm
Width (Metric) Printable Area 25.4 mm
Height (English) 3.75 in
Height (Metric) 95.3 mm
Height (Metric) Printable Area 25.4 mm
Alternate Part Number THT-10-427-3-YL