Learn More
Brady™ B-427 Self-laminating Vinyl Thermal Transfer Labels
Product Code. 11524314 Shop All Brady Products
Change view
Click to view available options
Color:
White, Transparent
Yellow
Height (Metric):
20 mm
38.1 mm
Vertical Repeat (Metric):
26 mm
41.3 mm
Width (Metric):
20 mm
38.1 mm
Unit Size:
1 roll
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (Metric) Vertical Repeat (Metric) Width (Metric) Unit Size
11524314 Yellow 20 mm 26 mm 20 mm 1 roll
11579903 White, Transparent 38.1 mm 41.3 mm 38.1 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11524314 Supplier Brady™ Supplier No. 800538
  • ONLINE EXCLUSIVE PRICE
    900.00€
    758.19€ / 1 rolls
    Save 141.81€
    15% Off
Estimated Shipment: 18-12-2026
Printable, self-laminating vinyl labels for Thermal Transfer Printers.
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
Specifications
Certifications/Compliance UL Recognized
Adhesive Type Acrylic, Permanent
Color Yellow
Finish Matte
Format Die-cut
Material Self-laminating Vinyl
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 76 mm Core Printers
Labels per Row 1
Quantity 1000 Labels
Shape Square
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 26 mm
Dimensions (L x W) 20.00 mm (W) x 20.00 mm (H)
Length (English) 0.788 in.
Width (English) 0.787 in.
Width (Metric) 20 mm
Width (Metric) Printable Area 20 mm
Height (English) 0.788 in.
Height (Metric) 20 mm
Alternate Part Number BPTLAB-35-427-YL