Learn More
Synergy Healthcare™ Azo™ Gusseted Reels
Product Code. 15651974
Change view
Click to view available options
Dimensions (L x W):
100 m x 100 mm
100 m x 150 mm
100 m x 200 mm
100 m x 300 mm
Quantity:
1 Ea.
4/Cs.
Unit Size:
Each
Pack of 2
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
15651974 100 m x 200 mm 4/Cs. Pack of 2
15631974 100 m x 100 mm 1 Ea. Pack of 4
15604977 100 m x 300 mm 4/Cs. Each
15641974 100 m x 150 mm 4/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15651974 Supplier Synergy Healthcare™ Supplier No. MRF1062
  • ONLINE EXCLUSIVE PRICE
    179.00€
    156.00€ / Pack of 2
    Save 23.00€
    12% Off
 Restricted Item
Estimated Shipment: 14-12-2026
High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Gusseted Reels are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.
  • Protects its contents from deterioration, damage and contamination
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Autoclavable Autoclavable
Certifications/Compliance EN868-5 and ISO11607-1
Color Blue
Sterility Non-sterile
Quantity 4/Cs.
Sterilization Method Steam and EO
Dimensions (L x W) 100 m x 200 mm
Product Type Sterilization Bag
Length (Metric) 100 m