Learn More
Synergy Healthcare™ Azo™ Gusseted Reels
Product Code. 15631974
Change view
Click to view available options
Dimensions (L x W):
100 m x 100 mm
100 m x 150 mm
100 m x 200 mm
100 m x 300 mm
Quantity:
1 Ea.
4/Cs.
Unit Size:
Each
Pack of 2
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity unitSize
15631974 100 m x 100 mm 1 Ea. Pack of 4
15604977 100 m x 300 mm 4/Cs. Each
15641974 100 m x 150 mm 4/Cs. Pack of 4
15651974 100 m x 200 mm 4/Cs. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15631974 Supplier Synergy Healthcare™ Supplier No. MRF1060
  • ONLINE EXCLUSIVE PRICE
    284.00€
    242.82€ / Pack of 4
    Save 41.18€
    14% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Gusseted Reels are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.
  • Protects its contents from deterioration, damage and contamination
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Autoclavable Autoclavable
Certifications/Compliance EN868-5 and ISO11607-1
Color Blue
Material Polyester, Polypropylene
Sterility Non-sterile
Quantity 1 Ea.
Sterilization Method Steam and EO
Dimensions (L x W) 100 m x 100 mm
Product Type Sterilization Reel
Length (Metric) 200 m