Learn More
Azlon™ Spare Stopcock for HDPE Aspirator
Replaces leaking or worn out stopcocks. Azlon™ Spare Stopcock for HDPE Aspirators create an excellent seal and are leakproof when installed properly.
Brand: Azlon™ SCBNP01
21.57 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.03000kg
Description
- For use with Heavy Duty Polypropylene (HDPE) Aspirators
- Opens and closes in a quarter turn
- Leakproof
Specifications
Spare Stopcock | |
HDPE | |
HDPE aspirator bottle |