Learn More
Azlon™ HDPE, Jerry Can
Click to view available options
:
Capacity: 25L
Description
- Lightweight and robust
- Corrosion resistant
- Suitable for food contact applications in accordance with EU and U.S. FDA regulations
Specifications
Specifications
| Capacity (English) | 6.6 gal. |
| Capacity (Metric) | 25 L |
| Dimensions (L x W x H) | 294 x 245 x 470mm |
| Material | HDPE |
| No. per Pack | 1 Ea. |
| Product Type | Jerry Can |