Learn More
Azlon™ PTFE Stirrer Bars
Product Code. 15778087
Click to view available options
Length (Metric):
75 mm
108 mm
159 mm
Diameter (Metric):
13 mm
27 mm
Unit Size:
Pack of 2
Product Code. 15778087
Brand: Azlon SWN677
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for use with large vessels.
  • Similar to cylindrical stirrer bars but give greater turbulence at low speed
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 13 mm
Length (Metric) 75 mm
Material Magnet
Quantity 2